missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10466082
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10466082 10 g 10g
10120882 1 g 1g
11306875 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10466082 Supplier Thermo Scientific Maybridge Supplier No. GK01821EA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-27-6
Molecular Formula C7H4ClF3N2O2
Molecular Weight (g/mol) 240.57
MDL Number MFCD00068146
InChI Key VLUBJYUOPNGBOQ-UHFFFAOYSA-N
Synonym methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate, methylchlorotrifluoromethylpyrimidinecarboxylate, methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, acmc-20aifi, pubchem10174, 2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine, methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca, 2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine, 2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
PubChem CID 3696469
IUPAC Name methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES COC(=O)C1=CN=C(Cl)N=C1C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 240.57
Percent Purity 97%
Chemical Name or Material Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.