missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-isothiocyanatothiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10059131
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10059131 1 g Amber Glass Bottle 1g
10762483 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10059131 Supplier Thermo Scientific Maybridge Supplier No. GK00289DA
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 81321-10-0
Molecular Formula C7H5NO2S2
Molecular Weight (g/mol) 199.24
MDL Number MFCD00052589
InChI Key RKBLURUDWDTMOB-UHFFFAOYSA-N
Synonym methyl 3-isothiocyanato-2-thiophenecarboxylate, 3-isothiocyanato-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-isothiocyanato-, methyl ester, methyl 3-isoisothiocyanatothiophene-2-carboxylate
PubChem CID 2777461
IUPAC Name methyl 3-isothiocyanatothiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)N=C=S

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material Methyl 3-isothiocyanatothiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.