missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylglyoxal Antibody (9E7), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59368PECY55
This item is not returnable.
View return policy
Description
Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Flow Cytometry.
Specifications
| Methylglyoxal | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human, Non-species specific | |
| IgG2a |
| Flow Cytometry | |
| 90000000 | |
| Flow Cytometry | |
| 2-oxopropanal, Pyruvaldehyde | |
| Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
| 0.1 mL | |
| Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins. | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction