missing translation for 'onlineSavingsMsg'
Learn More

Methylprednisolone 21-Propionate, TRC

Product Code. 30011694
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011694 2.5 g 2.5g
30005386 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011694 Supplier Toronto Research Chemicals Supplier No. TRCM3282302.5G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138804-88-3
Molecular Formula C25 H34 O6
Molecular Weight (g/mol) 430.53
Synonym (6α,11β)-11,17-Dihydroxy-6-methyl-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione
IUPAC Name [2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxo-ethyl] propanoate
SMILES CCC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3C[C@H](C)C4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C25H34O6/c1-5-21(29)31-13-20(28)25(30)9-7-17-16-10-14(2)18-11-15(26)6-8-23(18,3)22(16)19(27)12-24(17,25)4/h6,8,11,14,16-17,19,22,27,30H,5,7,9-10,12-13H2,1-4H3/t14-,16-,17-,19-,22+,23-,24-,25-/m0/s1
Formula Weight 430.2355
Percent Purity >95%
Chemical Name or Material Methylprednisolone 21-Propionate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.