missing translation for 'onlineSavingsMsg'
Learn More

Methyltin trichloride, 97%

Product Code. 11334849
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334849 1 g 1g
11324849 5 g 5g
11344849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334849 Supplier Thermo Scientific Alfa Aesar Supplier No. 071156.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 993-16-8 | CH3Cl3Sn | 240.095 g/mol

A chemical used in the synthesis of metallasilesesquiloxanes. Also used in the preparation of organotin(IV) complexes showing antibacterial activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 993-16-8
Molecular Formula CH3Cl3Sn
Molecular Weight (g/mol) 240.095
MDL Number MFCD00000514
InChI Key YFRLQYJXUZRYDN-UHFFFAOYSA-K
Synonym methyltin trichloride, methyltrichlorotin, stannane, trichloromethyl, trichloromethyltin, monomethyltin trichloride, trichloro methyl stannane, methyltrichlorostannane, tin, methyl-, trichloride, unii-q2e6v14u06, ccris 6327
PubChem CID 101940
IUPAC Name trichloro(methyl)stannane
SMILES C[Sn](Cl)(Cl)Cl

Specifications

Melting Point 43°C
Quantity 1 g
UN Number UN2923
Sensitivity Moisture sensitive
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 240.11
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Methyltin trichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.