missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylarsonium iodide, 98%, Thermo Scientific™

Product Code. 11305229
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305229 1 g 1g
11315229 5 g 5g
11325229 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11305229 Supplier Thermo Scientific Alfa Aesar Supplier No. A18599.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-33-8
Molecular Formula C19H18AsI
Molecular Weight (g/mol) 448.179
MDL Number MFCD00031543
InChI Key GHLKOMSNOFJXDT-UHFFFAOYSA-M
Synonym methyltriphenylarsonium iodide, arsonium, methyltriphenyl-, iodide, methyltriphenylarsanium iodide, methyl triphenylarsonium iodide, arsonium, methyltriphenyl-, iodide 1:1, c19h18as.i, acmc-209d3h, triphenylmethylarsonium iodide, methyl triphenyl arsanium iodide, arsonium,methyltriphenyl-, iodide 1:1
PubChem CID 73911
IUPAC Name methyl(triphenyl)arsanium;iodide
SMILES C[As+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]

Specifications

Melting Point 173°C to 177°C
Quantity 1 g
UN Number UN3465
Beilstein 3639838
Sensitivity Light sensitive
Formula Weight 448.18
Percent Purity 98%
Chemical Name or Material Methyltriphenylarsonium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.