missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium iodide, 98%

Product Code. 11487547
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11487547

Brand: Thermo Scientific Alfa Aesar A15644.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2065-66-9 | C19H18IP | 404.23 g/mol

Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

Solubility
Soluble in water, acetone, dichloromethane , methanol.

Notes
Light sensitive and hygroscopic. Store in dark. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2065-66-9
Molecular Formula C19H18IP
Molecular Weight (g/mol) 404.23
MDL Number MFCD00066175
InChI Key JNMIXMFEVJHFNY-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium iodide, methyl triphenylphosphonium iodide, methyltriphenylphosphanium iodide, methyl triphenyl phosphonium iodide, phosphonium, methyltriphenyl-, iodide, methyl triphenylphosphoniumiodide, methyl triphenyl phosphanium iodide, triphenylmethyl phosphonium iodide, pubchem15730
PubChem CID 638159
IUPAC Name methyl(triphenyl)phosphanium;iodide
SMILES [I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 185°C to 188°C
Odor Odorless
Quantity 25 g
UN Number UN3464
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water,acetone,dichloromethane ,methanol.
Formula Weight 404.23
Percent Purity 98%
Chemical Name or Material Methyltriphenylphosphonium iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.