missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium iodide, 98%

Product Code. 11497547
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497547 100 g 100g
11487547 25 g 25g
11407557 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497547 Supplier Thermo Scientific Alfa Aesar Supplier No. A15644.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2065-66-9 | C19H18IP | 404.23 g/mol

Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

Solubility
Soluble in water, acetone, dichloromethane , methanol.

Notes
Light sensitive and hygroscopic. Store in dark. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2065-66-9
Molecular Formula C19H18IP
Molecular Weight (g/mol) 404.23
MDL Number MFCD00066175
InChI Key JNMIXMFEVJHFNY-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium iodide, methyl triphenylphosphonium iodide, methyltriphenylphosphanium iodide, methyl triphenyl phosphonium iodide, phosphonium, methyltriphenyl-, iodide, methyl triphenylphosphoniumiodide, methyl triphenyl phosphanium iodide, triphenylmethyl phosphonium iodide, pubchem15730
PubChem CID 638159
IUPAC Name methyl(triphenyl)phosphanium;iodide
SMILES [I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 185°C to 188°C
Odor Odorless
Quantity 100 g
UN Number UN3464
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water,acetone,dichloromethane ,methanol.
Formula Weight 404.23
Percent Purity 98%
Chemical Name or Material Methyltriphenylphosphonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.