missing translation for 'onlineSavingsMsg'
Learn More

Metoclopramide hydrochloride, 98%

Product Code. 16554344 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16554344 5 g 5g
16564344 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16554344 Supplier Thermo Scientific Acros Supplier No. 461820050
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7232-21-5 | C14H23Cl2N3O2 | 336.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7232-21-5
Molecular Formula C14H23Cl2N3O2
Molecular Weight (g/mol) 336.26
InChI Key RVFUNJWWXKCWNS-UHFFFAOYSA-N
IUPAC Name 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride
SMILES Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC

Specifications

Melting Point 171.0°C to 173.0°C
Assay Percent Range 98%
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: soluble in methanol
Formula Weight 336.26
Chemical Name or Material Metoclopramide hydrochloride
compliance-icons
Product Identifier
  • Metoclopramide hydrochloride, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide monohydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.