Learn More
Metro™ High-Density qwikTRAK™ Track Sets
Create tracks up to 21 ft.
530.00 € - 2135.00 €
Specifications
For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system |
---|---|
Product Type | Track Set |
Product Code | Brand | Depth (English) | Depth (Metric) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (English) | Depth (Metric) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
15926881
|
Metro™
BTS10NA |
120 in. | 304.8 cm | 14 lb. | 6.4 kg |
1060.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15936881
|
Metro™
BTS11NA |
132 in. | 335.28 cm | 15 lb. | 6.8 kg |
1145.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15946881
|
Metro™
BTS12NA |
144 in. | 365.76 cm | 17 lb. | 7.7 kg |
1220.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15956881
|
Metro™
BTS13NA |
156 in. | 396.24 cm | 18 lb. | 8.2 kg |
1300.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15966881
|
Metro™
BTS14NA |
168 in. | 426.72 cm | 19 lb. | 8.6 kg |
1375.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15976881
|
Metro™
BTS15NA |
180 in. | 457.2 cm | 21 lb. | 9.5 kg |
1670.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15996881
|
Metro™
BTS17NA |
204 in. | 518.16 cm | 23 lb. | 10.5 kg |
1825.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15986881
|
Metro™
BTS16NA |
192 in. | 487.68 cm | 22 lb. | 10 kg |
1745.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15906891
|
Metro™
BTS18NA |
216 in. | 548.64 cm | 25 lb. | 11.4 kg |
1905.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15916891
|
Metro™
BTS19NA |
228 in. | 579.12 cm | 26 lb. | 12 kg |
1980.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15936891
|
Metro™
BTS20NA |
240 in. | 609.6 cm | 27 lb. | 12.3 kg |
2060.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15946891
|
Metro™
BTS21NA |
252 in. | 640.08 cm | 29 lb. | 13.2 kg |
2135.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15926891
|
Metro™
BTS2.5NA |
30 in. | 76.2 cm | 4 lb. | 1.8 kg |
530.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15976891
|
Metro™
BTS8NA |
96 in. | 243.84 cm | 11 lb. | 5 kg |
910.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15986891
|
Metro™
BTS9NA |
108 in. | 274.32 cm | 13 lb. | 5 kg |
980.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15956891
|
Metro™
BTS6NA |
72 in. | 182.88 cm | 9 lb. | 4 kg |
545.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15966891
|
Metro™
BTS7NA |
84 in. | 213.36 cm | 10 lb. | 4 kg |
620.00 €
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q and MetroMax i shelving systems
- Sets include tracks and hardware needed to assemble system up to 21 ft. (640cm)
- 17-823-715 (BTS2.5NA) includes a joining kit to connect the track to other track sets
Specifications
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system | |
Track Set |