missing translation for 'onlineSavingsMsg'
Learn More

Metsulfovax, Dr. Ehrenstorfer

Product Code. 30309936
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309936 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309936 Supplier Dr. Ehrenstorfer Supplier No. DREC15208000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metsulfovax
CAS 21452-18-6
Analyte or Component Names Metsulfovax
Shipping Condition Room Temperature
Molecular Formula C12 H12 N2 O S
Certifications/Compliance ISO/IEC 17025
Synonym 5-Thiazolecarboxanilide, 2,4-dimethyl- (8CI), 2,4-Dimethyl-N-phenyl-5-thiazolecarboxamide, 2,4-Dimethyl-5-carboxanilidothiazole, 2,4-Dimethyl-N-phenyl-1,3-thiazole-5-carboxamide, 2,4-Dimethylthiazole-5-carboxanilide, ALG, DCT, DCT (2,4-dimethyl-5-carboxanilidothiazole), G 696, Metsulfovax, Provax, Provax (fungicide)
InChI Formula InChI=1S/C12H12N2OS/c1-8-11(16-9(2)13-8)12(15)14-10-6-4-3-5-7-10/h3-7H,1-2H3,(H,14,15)
Solution Type Neat
SMILES Cc1nc(C)c(s1)C(=O)Nc2ccccc2
IUPAC Name 2,4-dimethyl-N-phenyl-1,3-thiazole-5-carboxamide
Industry Type Environmental Testing
Molecular Weight (g/mol) 232.3
Quantity 25 mg
Formula Weight 232.067 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.