missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | METTL1 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657648
|
Novus Biologicals
NBP2-99811-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641987
|
Novus Biologicals
NBP2-99811-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
METTL1 Polyclonal antibody specifically detects METTL1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| METTL1 | |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 4234 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w | |
| Produced in rabbits immunized with purified, recombinant Human METTL1 (Accession#: NP_005362.3; Asp 32-Gln 265) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title