missing translation for 'onlineSavingsMsg'
Learn More

METTL10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

127.00 € - 387.00 €

Specifications

Antigen METTL10
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Applications Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18636264
View Documents
Novus Biologicals
NBP3-21446-25ul
25 μg
127.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18654574
View Documents
Novus Biologicals
NBP3-21446-100ul
100 μg
387.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

METTL10 Polyclonal antibody specifically detects METTL10 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

METTL10
Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
C10orf138, chromosome 10 open reading frame 138, EC 2.1.1.-, Em:AC068896.3, FLJ13019, methyltransferase like 10, methyltransferase-like protein 10
Produced in rabbits immunized with E. coli-derived Human METTL10 fragment
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
PBS, pH 7.0
399818
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.