missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48804-25ul
This item is not returnable.
View return policy
Description
METTL26 Polyclonal antibody specifically detects METTL26 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| METTL26 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| chromosome 16 open reading frame 13, hypothetical protein LOC84326, JFP2, MGC13114 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AERNKDPILHVLRQYLDPAQRGVRVLEVASGSGQHAAHFARAFPLAEWQPSDVDQRCLDSIAATTQAQGLTNVKAPLHLDVTWGWEHWGGILPQSLDLLLC | |
| 25 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84326 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction