missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™

Product Code. 10166380
Change view
Click to view available options
Quantity:
20 mL
Conductivity:
12.88 mS/cm
1413 μS/cm
84 μS/cm
Unit Size:
600mL
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conductivity Unit Size
10166380 20 mL 1413 μS/cm 600mL
10528632 20 mL 12.88 mS/cm 600mL
15574843 20 mL 84 μS/cm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10166380 Supplier Mettler Toledo Online Supplier No. 51302049
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recommended NIST traceable solutions for calibrating or verifying METTLER TOLEDO conductivity electrodes.

  • For daily verification: to monitor health of electrode and ensure measurement accuracy
  • For initial calibration of nominal cell constant electrodes
  • Be sure to screw cap tightly immediately after use (sachets minimize the chance of error due to environmental interaction)
  • Conductivity standards subject to change values when exposed to environment; water evaporation raises conductivity values
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Chloride Solution
Packaging 30 x 20 mL
CAS 7447-40-7, 7732-18-5
Traceability to NIST Yes
Conductivity 1413 μS/cm
Molecular Formula KCl
Color Blue/White
pH Approx. 6
Melting Point 0°C
Boiling Point 100°C
For Use With (Equipment) Conductivity Electrodes
Solution Type Conductivity Standard
Type Conductivity Standard
Industry Type Calibration of Conductivity Meter
Quantity 20 mL
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.