Learn More
METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters
Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.
Brand: METTLER TOLEDO™ 30207956
Code : NEW
Additional Details : CAS Number : 7447-40-7 Weight : 2.84000kg
Includes: Instrument, electrode clip, InLab 738-ISM and sachets of conductivity standards (1413µS/cm and 12.88mS/cm), bottles for calibration, operating instructions, quick guide, test report declaration of conformity, uGo™ carrying case and batteries
Description
- Dust proof
- Water proof
- IP67 Rated
- pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
- Compatible with a wide range of IP67-rated electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Data storage available; capacity dependent on model
- Export data to a printer or Excel file
- EasyDirect software available for data transfer (model dependent)
- Rubber holster available for single-handed measurements
- uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
Certifications
ISO 9001, ISO 14001
Specifications
±0.5% | |
±0.5% | |
4 x 1.5 V AA or 1.3 V NiMH acc. | |
Manual entry | |
LCD | |
0.01 to 1 (auto-range) | |
0.00 to 42 psu | |
-5°C to +105°C | |
InLab 738-ISM conductivity/temperature electrode; uGo carrying/protective case | |
Single channel; conductivity, TDS, salinity | |
0.01 to 1 (auto-range) | |
0.01 to 1 | |
222 x 70 x 35 mm |
±0.5% | |
250 to 400 hr. | |
1 | |
ISO 9001, ISO 14001 | |
IP 67 | |
0.00 to 100.0 Mohm/cm | |
0.01 mg/L to 300 g/L | |
For aqueous samples with mid- to high conductivity; ideal for measurements on the go. | |
LTW 7-pin (sensor) | |
Seven2Go S3 Conductivity Meter: Field Kit | |
0.01 to 0.1 | |
Automatic or Manual | |
270 g |