missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences MEXi-293E Cells

IBA Lifesciences GmbH MEXi-293E Cells are an efficient and cost effective mammalian protein expression system.

Brand:  IBA Lifesciences 2-6001-001

Additional Details : Weight : 0.00150kg

Product Code. 15513087

  • 432.00 € / 1.50mL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MEXi-293E cells are human embryonic kidney (HEK) cells derived from the 293 cell line and used for propagation of plasmids and expression of recombinant proteins in mammalian cells.
The cell line expresses the EBNA-1 protein for episomal replication of oriP-harboring plasmids (pDSG-IBA vectors), is adapted for growth in suspension, and for use with the MEXi cell culture medium. The MEXi system was developed to offer an affordable mammalian protein expression system coming along with all essential components:
• MEXi-293E cell line
• A variety of pDSG expression vectors
• MEXi-CM culture medium
• MEXi-TM transfection medium Further advantages:
• Transient expression for time- and cost-efficient protein production
• Multiple expression vectors with different features
• Optimized for Twin-Strep-tag™:Strep-Tactin™XT system for highly pure proteins
• IP-free system
Specifications

Specifications

Human Embryonic Kidney (HEK) Cells
1.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers