missing translation for 'onlineSavingsMsg'
Learn More

Mexiletine hydrochloride, MedChemExpress

Product Code. 16478026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478026 100 mg 100mg
16431986 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478026 Supplier Medchem Express Supplier No. HYA0093
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mexiletine hydrochloride (KOE-1173 hydrochloride), a Class IB antianhythmic, is a non-selective voltage-gated sodium channel blocker.

Mexiletine hydrochloride (KOE-1173 hydrochloride), a Class IB antianhythmic, is a non-selective voltage-gated sodium channel blocker.

Specifications

Chemical Name or Material Mexiletine hydrochloride
CAS 1-4-5370
Color White
Purity Grade Notes Research
Molecular Formula C11H18ClNO
Quantity 100 mg
Synonym KOE-1173 hydrochloride
Solubility Information H2O : 100 mg/mL (463.56 mM; Need ultrasonic) ∣DMSO : ≥ 41 mg/mL (190.06 mM)
SMILES CC(N)COC1=C(C)C=CC=C1C.[H]Cl
Molecular Weight (g/mol) 215.72
Formula Weight 215.72
Percent Purity 98.83%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.