missing translation for 'onlineSavingsMsg'
Learn More

MFF Antibody (SR1564), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00 € - 449.00 €

Specifications

Antigen MFF
Clone SR1564
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18653855
View Documents
Novus Biologicals
NBP3-22301-100ul
100 μg
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18693144
View Documents
Novus Biologicals
NBP3-22301-50ul
50 μg
338.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MFF Monoclonal antibody specifically detects MFF in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

MFF
Western Blot 1:500-1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
mitochondrial fission factor
A synthesized peptide derived from human MFF (Uniprot #: Q9GZY8)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1564
Western Blot
Unconjugated
Rabbit
Apoptosis, Cell Biology, Endocrinology
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
56947
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.