Learn More
MFSD2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30555-1-AP-20UL
Description
The MFSD2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MFSD2. This antibody recognizes human, mouse, rat antigen. The MFSD2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| MFSD2 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| FLJ14490, FLJ35904, FP1147, HMFN0656, MFSD2, MFSD2A, PP10484, PP9177, RP3 342P20.1, UNQ300/PRO341 | |
| 20 μL | |
| 84879 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |