missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGAT5 Recombinant Rabbit Monoclonal Antibody (7W10I10)
Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Specifications
| Antigen | MGAT5 |
|---|---|
| Clone | 7W10I10 |
| Concentration | 0.55 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30282576
|
Invitrogen™
MA557861 |
20 μL |
176.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30284462
|
Invitrogen™
MA557862 |
100 μL |
436.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: CHGQVMWPPL SALQVKLAEP GQSCKQVCQE SQLICEPSFF QHLNKDKDML KYKVTCQSSE LAKDILVPSF DPKNKHCVFQ GDLLLFSCAG AHPRHQRVCP CRDFIKGQVA LCKDCL.
Catalyzes the addition of N-acetylglucosamine in beta 1- 6 linkage to the alpha-linked mannose of biantennary N-linked oligosaccharides. It is one of the most important enzymes involved in the regulation of the biosynthesis of glycoprotein oligosaccharides.Specifications
| MGAT5 | |
| 0.55 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q08834, Q09328, Q8R4G6 | |
| 107895, 4249, 65271 | |
| Recombinant protein (or fragment). | |
| Antibody |
| 7W10I10 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| MGAT5 | |
| 4930471A21Rik; 5330407H02Rik; AI480971; alpha-1,6-mannosyl-glycoprotein 6-beta-N-acetylglucosaminyltransferase; alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase V; beta1,6N-acetylglucosaminyltransferase V; EC 2.4.1.155; GGNT5; GlcNAc-T V; GlcNAc-TV; GNT-V; GNT-VA; GNT-VGNT-VA; mannoside acetylglucosaminyltransferase 5; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase; Mgat5; N-acetylglucosaminyltransferase V; N-acetylglucosaminyl-transferase V; Secreted alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; Secreted beta-1,6-N-acetylglucosaminyltransferase V; Secreted GNT-V | |
| MGAT5 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title