missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGP Antibody (8C3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42999
This item is not returnable.
View return policy
Description
MGP Monoclonal antibody specifically detects MGP in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| MGP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4256 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Immunohistochemistry, Western Blot | |
| 8C3 | |
| Western Blot 1:500 - 1:5000, ELISA, Immunohistochemistry 1:500 - 1:1500 | |
| Cell growth-inhibiting gene 36 protein, matrix Gla protein, MGLAP, NTI | |
| matrix Gla protein - (Uniprot# P08493) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction