Learn More
Invitrogen™ MGST1 Recombinant Rabbit Monoclonal Antibody (24GB5110)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557077
 View more versions of this product
Product Code. 30275514
  • 500.00 € / 100µL
Description
MGST1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MGST1
Recombinant Monoclonal
Unconjugated
MGST1
1500002K10Rik; glutathione S-transferase; glutathione S-transferase 12; Gst; GST12; MGC14525; MGST; mGST1; MGST-I; Microsomal glutathione S-transferase 1; microsomal glutathione S-transferase 1-like protein; microsomal glutathione-S-transferase; microsomal GST-1; Microsomal GST-I
Rabbit
Affinity chromatography
RUO
4257, 56615
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5110
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P10620, Q91VS7
MGST1
A synthesized peptide derived from human MGST1 (aa 20-55).
100 μL
Primary
Human, Mouse