Learn More
MHC Class II Antibody (OX-6) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43539
Product Code. 30279785
  • 615.00 € / 0.20mg
Description
MHC Class II Monoclonal antibody specifically detects MHC Class II in Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Radioimmune Assays,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MHC Class II
Monoclonal
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmunoassay
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
This antibody was prepared by subcutaneously injecting non-polymorphic MHC class II (Ia-like glycoproteins) from the membranes of rat thymocytes into BALB/c mice.
0.2 mg
Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
3108
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Radioimmune Assays (RIA), Immunocytochemistry/Immunofluorescence
OX-6
Unconjugated
PBS
Rabbit
Protein A purified
RUO
Primary
Mouse, Rat
Purified