missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MHC Class II I-Ab Monoclonal Antibody (AF6-120.1), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | MHC Class II I-Ab |
|---|---|
| Clone | AF6-120.1 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055682
|
Invitrogen™
M046T03Y04-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054996
|
Invitrogen™
M046T02Y04-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MHC Class II I-Ab Monoclonal antibody specifically detects MHC Class II I-Ab in Mouse samples. It is validated for Flow CytometrySpecifications
| MHC Class II I-Ab | |
| 0.1 mg/mL | |
| Monoclonal | |
| Mouse | |
| Mouse | |
| P01921, P06342, P06343, P06344, P06345, P06346, P14483 | |
| 14961 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| AF6-120.1 | |
| Flow Cytometry | |
| NovaFluor Yellow 660 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Abeta; AI845868; CELIAC1; DADB-249P12.2; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2Eb; H-2Eb; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; HLA-DQB; Ia2; Ia-2; Ia4; Ia-4; IAb; I-Abeta; IDDM1; major histocompatibility complex class II beta chain; MGC163794; MGC163796; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 | |
| IgG2a κ | |
| Affinity chromatography | |
| H2-Ab1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title