missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Rat, APC, Clone: M5/114.15.2, enQuireBio™
Rat Monoclonal Antibody
131.25 € - 288.75 €
Specifications
| Antigen | MHC Class II |
|---|---|
| Clone | M5/114.15.2 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15992378
|
EnquireBio
QAB70-APC-25ug |
25 μg |
131.25 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15982378
|
EnquireBio
QAB70-APC-100ug |
100 μg |
288.75 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| MHC Class II | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 14969 | |
| IgG2b κ | |
| Protein A/G Purified |
| M5/114.15.2 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| NP_034512 | |
| H2-Ab1 and H2-Eb1 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title