missing translation for 'onlineSavingsMsg'
Learn More

MICAL3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00 € - 572.00 €

Specifications

Antigen MICAL3
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18292054
View Documents
Novus Biologicals
NBP2-56826
100 μL
572.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18665938
View Documents
Novus Biologicals
NBP2-56826-25ul
25 μL
280.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MICAL3 Polyclonal specifically detects MICAL3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MICAL3
Polyclonal
Rabbit
Human
microtubule associated monoxygenase, calponin and LIM domain containing 3
MICAL3
IgG
Affinity Purified
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
57553
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MELASGAPRPRKASSAPSQGKERRPDSPTRPTLRGSEEPTLKHEATSEEVLSPPSDSGGPDGSFTSSEGSSGKSKKRSSLFSPRRNKKEKK
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.