missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Michigan Pesticide Mixture 2 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenthrin∣Boscalid∣Cyfluthrin∣Cypermethrin∣Fenoxycarb∣Fipronil∣Flonicamid∣Fludioxonil∣Imazalil∣Imidacloprid∣Methiocarb∣Myclobutanil∣Permethrin∣Prallethrin∣Pyrethrins∣Spinosad∣Thiacloprid∣Trifloxystrobin |
| CAS | 71751-41-2, 135410-20-7, 116-06-3, 131860-33-8, 82657-04-3, 188425-85-6 |
| Analyte or Component Names | Abamectin∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenthrin∣Boscalid∣Cyfluthrin∣Cypermethrin∣Fenoxycarb∣Fipronil∣Flonicamid∣Fludioxonil∣Imazalil∣Imidacloprid∣Methiocarb∣Myclobutanil∣Permethrin∣Prallethrin∣Pyrethrins∣Spinosad∣Thiacloprid∣Trifloxystrobin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Industry Type | Food Testing |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 100 μg/mL∣Acetamiprid 100 μg/mL∣Aldicarb 100 μg/mL∣Azoxystrobin 100 μg/mL∣Bifenthrin 100 μg/mL∣Boscalid 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Fenoxycarb 100 μg/mL∣Fipronil 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL∣Imazalil 100 μg/mL∣Imidacloprid 100 μg/mL∣Methiocarb 100 μg/mL∣Myclobutanil 100 μg/mL∣Permethrin 100 μg/mL∣Prallethrin 100 μg/mL∣Pyrethrins 100 μg/mL∣Spinosad 100 μg/mL∣Thiacloprid 100 μg/mL∣Trifloxystrobin 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?