missing translation for 'onlineSavingsMsg'
Learn More

Michigan Pesticide Mixture 2 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316784
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316784 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316784 Supplier Dr. Ehrenstorfer Supplier No. DREA50000100AL
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abamectin∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenthrin∣Boscalid∣Cyfluthrin∣Cypermethrin∣Fenoxycarb∣Fipronil∣Flonicamid∣Fludioxonil∣Imazalil∣Imidacloprid∣Methiocarb∣Myclobutanil∣Permethrin∣Prallethrin∣Pyrethrins∣Spinosad∣Thiacloprid∣Trifloxystrobin
CAS 71751-41-2, 135410-20-7, 116-06-3, 131860-33-8, 82657-04-3, 188425-85-6
Analyte or Component Names Abamectin∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenthrin∣Boscalid∣Cyfluthrin∣Cypermethrin∣Fenoxycarb∣Fipronil∣Flonicamid∣Fludioxonil∣Imazalil∣Imidacloprid∣Methiocarb∣Myclobutanil∣Permethrin∣Prallethrin∣Pyrethrins∣Spinosad∣Thiacloprid∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Abamectin 100 μg/mL∣Acetamiprid 100 μg/mL∣Aldicarb 100 μg/mL∣Azoxystrobin 100 μg/mL∣Bifenthrin 100 μg/mL∣Boscalid 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Fenoxycarb 100 μg/mL∣Fipronil 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL∣Imazalil 100 μg/mL∣Imidacloprid 100 μg/mL∣Methiocarb 100 μg/mL∣Myclobutanil 100 μg/mL∣Permethrin 100 μg/mL∣Prallethrin 100 μg/mL∣Pyrethrins 100 μg/mL∣Spinosad 100 μg/mL∣Thiacloprid 100 μg/mL∣Trifloxystrobin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.