missing translation for 'onlineSavingsMsg'
Learn More

Miconazole, 97%, Thermo Scientific™

Inhibits cytochrome P450-dependent 14-alpha-demethylase

Brand:  Thermo Scientific Alfa Aesar J60872.03

Quantity 1g

 View more versions of this product

Product Code. 15473569

  • ONLINE EXCLUSIVE PRICE
    34.80 €
    34.10 € / 1g
    Save 0.70 €
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Freely soluble in alcohols or acetone. Also soluble in DMF or chloroform. Insoluble in water

Notes
Protect from light. Store in well-closed containers.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

22916-47-8
416.12
BYBLEWFAAKGYCD-UHFFFAOYNA-N
4189
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
C18H14Cl4N2O
MFCD00216019
miconazole, monistat, monistat iv, daktarin iv, miconazolo, miconazolum, miconazol, minostate, dactarin, monistat-derm
CHEBI:82892
ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

22916-47-8
MFCD00216019
Light sensitive
miconazole, monistat, monistat iv, daktarin iv, miconazolo, miconazolum, miconazol, minostate, dactarin, monistat-derm
BYBLEWFAAKGYCD-UHFFFAOYNA-N
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
4189
416.13
Miconazole
C18H14Cl4N2O
1g
14,6178
Freely soluble in alcohols or acetone; Also soluble in DMF or chloroform; Insoluble in water
ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
416.12
CHEBI:82892
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.