missing translation for 'onlineSavingsMsg'
Learn More

Miconazole, 97%, Thermo Scientific™

Inhibits cytochrome P450-dependent 14-alpha-demethylase

34.10 € - 185.00 €

Chemical Identifiers

CAS 22916-47-8
Molecular Formula C18H14Cl4N2O
Molecular Weight (g/mol) 416.12
MDL Number MFCD00216019
InChI Key BYBLEWFAAKGYCD-UHFFFAOYNA-N
Synonym miconazole, monistat, monistat iv, daktarin iv, miconazolo, miconazolum, miconazol, minostate, dactarin, monistat-derm
PubChem CID 4189
ChEBI CHEBI:82892
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
SMILES ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473569
SDS View Documents
Thermo Scientific Alfa Aesar
J60872.03
1g
ONLINE EXCLUSIVE PRICE
34.80 €
34.10 € / 1g
Save 0.70 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15483569
SDS View Documents
Thermo Scientific Alfa Aesar
J60872.06
5g
ONLINE EXCLUSIVE PRICE
53.40 €
52.31 € / 5g
Save 1.09 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15493569
SDS View Documents
Thermo Scientific Alfa Aesar
J60872.14
25g
ONLINE EXCLUSIVE PRICE
189.00 €
185.00 € / 25g
Save 4.00 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Freely soluble in alcohols or acetone. Also soluble in DMF or chloroform. Insoluble in water

Notes
Protect from light. Store in well-closed containers.
Specifications

Chemical Identifiers

22916-47-8
416.12
BYBLEWFAAKGYCD-UHFFFAOYNA-N
4189
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
C18H14Cl4N2O
MFCD00216019
miconazole, monistat, monistat iv, daktarin iv, miconazolo, miconazolum, miconazol, minostate, dactarin, monistat-derm
CHEBI:82892
ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

22916-47-8
MFCD00216019
14,6178
Freely soluble in alcohols or acetone; Also soluble in DMF or chloroform; Insoluble in water
ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
416.12
CHEBI:82892
99%
C18H14Cl4N2O
Light sensitive
miconazole, monistat, monistat iv, daktarin iv, miconazolo, miconazolum, miconazol, minostate, dactarin, monistat-derm
BYBLEWFAAKGYCD-UHFFFAOYNA-N
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
4189
416.13
Miconazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P285-P301+P312-P304+P341-P330-P342+P311-P501c

H302-H334

missing translation for 'einecsNumber' : 245-324-5

missing translation for 'rtecsNumber' : NI4770000

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.