missing translation for 'onlineSavingsMsg'
Learn More

Miconazole (Free Base), Fisher BioReagents

Product Code. 10386933
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10386933

Brand: Fisher BioReagents BP26681

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Miconazole is used as an antifungal agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22916-47-8
Molecular Formula C18H14Cl4N2O
Molecular Weight (g/mol) 416.12
MDL Number MFCD00216019
InChI Key BYBLEWFAAKGYCD-UHFFFAOYNA-N
Synonym 1-[2, 4-Dichloro-β-([2, 4-dichloro-benzyl]oxy)phenethyl]imidazole
PubChem CID 4189
ChEBI CHEBI:82892
SMILES ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

Type Miconazole
Physical Form Solid
Melting Point 135°C
Ignition Residue < 0.2%
Loss on Drying < 0.5%
Packaging Amber Glass
Color White
Quantity 1 g
Assay Percent Range 98%
Merck Index 15, 6257
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole
Formula Weight 416.13
Chemical Name or Material Miconazole, Free Base
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Miconazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.