missing translation for 'onlineSavingsMsg'
Learn More

Miconazole (Free Base), Fisher BioReagents

Product Code. 11468403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11468403 5 g Amber Glass 5g
10386933 1 g Amber Glass 1g
10596764 25 g Amber Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468403 Supplier Fisher BioReagents Supplier No. BP26685

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Miconazole is used as an antifungal agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22916-47-8
Molecular Formula C18H14Cl4N2O
Molecular Weight (g/mol) 416.12
MDL Number MFCD00216019
InChI Key BYBLEWFAAKGYCD-UHFFFAOYNA-N
Synonym 1-[2, 4-Dichloro-β-([2, 4-dichloro-benzyl]oxy)phenethyl]imidazole
PubChem CID 4189
ChEBI CHEBI:82892
SMILES ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

Type Miconazole
Physical Form Solid
Melting Point 135°C
Ignition Residue < 0.2%
Loss on Drying < 0.5%
Packaging Amber Glass
Color White
Quantity 5 g
Assay Percent Range 98%
Merck Index 15, 6257
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole
Formula Weight 416.13
Chemical Name or Material Miconazole, Free Base
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Miconazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.