missing translation for 'onlineSavingsMsg'
Learn More

Miconazole Free Base MP Biomedicals

Product Code. 11426321 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426321 5 g 5g
11416321 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426321 Supplier MP Biomedicals Supplier No. 0219391505
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 22916-47-8
Molecular Formula C18H14Cl4N2O
Molecular Weight (g/mol) 416.12
MDL Number MFCD00216019
InChI Key BYBLEWFAAKGYCD-UHFFFAOYNA-N
Synonym miconazole, monistat, monistat iv, daktarin iv, miconazolo, miconazolum, miconazol, minostate, dactarin, monistat-derm
PubChem CID 4189
ChEBI CHEBI:82892
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
SMILES ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

Quantity 5 g
Formula Weight 416.1
Chemical Name or Material Miconazole, Free Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.