missing translation for 'onlineSavingsMsg'
Learn More

(±)-Miconazole nitrate salt, Thermo Scientific Chemicals

Product Code. 15552223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15552223 25 g 25g
15532223 1 g 1g
15542223 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15552223

Brand: Thermo Scientific Acros 291830250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22832-87-7
Molecular Formula C18H15Cl4N3O4
Molecular Weight (g/mol) 479.14
MDL Number MFCD00058161
InChI Key MCCACAIVAXEFAL-UHFFFAOYNA-N
PubChem CID 68553
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

Type (±)-Miconazole Nitrate Salt
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C, 2 h)
Quantity 25 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 479.14
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material (±)-Miconazole nitrate salt
compliance-icons compliance-icons
Product Identifier
  • (±)-Miconazole nitrate salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.