missing translation for 'onlineSavingsMsg'
Learn More

Miconazole, TRC

Product Code. 30012525
Change view
Click to view available options
Quantity:
50 mg
2.5 g
10 g
Unit Size:
10g
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012525 2.5 g 2.5g
30007529 10 g 10g
30005536 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012525 Supplier Toronto Research Chemicals Supplier No. TRCM3425052.5G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22916-47-8
Molecular Formula C18 H14 Cl4 N2 O
Molecular Weight (g/mol) 416.13
Synonym Imidazole, 1-[2,4-dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl]- (8CI), 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole, (±)-Miconazole, Daktanol, Daktarin IV, Florid-F, Lauriad, Loramyc, MJR 1762, Miconazole, Monistat IV, NSC 170986, R 18134, Zimybase
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole
SMILES Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C18H14Cl4N2O/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22/h1-8,11,18H,9-10H2
Formula Weight 413.986
Percent Purity >95%
Chemical Name or Material Miconazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.