missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates

Product Code. 16834683 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
20 plates
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16834683 20 plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16834683 Supplier Applied Biosystems™ Supplier No. A36931
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs.

EnduraPlate™ plastic consumables are validated with Applied Biosystems™ instruments for optimal fit and performance. They are constructed to ANSI/SBS standards and are well-suited for automated workflows. MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates are skirted and fit 384-well (0.02 mL) thermal blocks.

  • Optimized for performance with Applied Biosystems™ instruments
  • Certified DNA-, RNase-, and PCR inhibitor-free
  • Constructed to ANSI/SBS standards for compatibility with high-throughput laboratories and robotic handling
  • Easy-to-read black well-identification text to minimize pipetting errors

EnduraPlate™ reaction plates use polypropylene for a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from the thermal block to the sample, while a polycarbonate frame provides mechanical stability, flatness, and durability that is compatible with automation/robotics. Easy-to-read, black well-identification text helps to minimize pipetting mistakes and improves sample preparation.

MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates are provided without barcodes, enabling customization with your own barcodes.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Store at room temperature.
Packaging 20 Plates
Product Line MicroAmp
Color Optical, Clear
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Skirted
Material Polycarbonate, Polypropylene
Volume (Metric) Well 40 μL
No. of Wells 384
Quantity 20 plates
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.