missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 32-Well Clear Adhesive Film

Product Code. 15845066 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 Films
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15845066 100 Films Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 15845066 Supplier Applied Biosystems™ Supplier No. A32812

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

For use with MicroAmp™ TriFlex 3 x 32-Well PCR Reaction Plate

  • Easy to apply and remove - simply peel off backing and apply firmly to 32-well plate
  • Provides an alternative to cap strips and seals 32-well samples securely to prevent sample evaporation and well-to-well contamination
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polyethylene
No. of Wells 32
Quantity 100 Films
For Use With (Equipment) MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate
Shipping Condition Room Temperature
Product Type Film, Seal, or Applicator
Product Line MicroAmp
Plate Compatibility 32-well Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.