Learn More
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates
Product Code. 10577091
Change view
Click to view available options
Binding Property:
High Binding
Medium Binding
Well Design:
Chimney Well
Standard Well
Unit Size:
Pack of 40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Binding Property Well Design Unit Size
10577091 High Binding Standard Well Pack of 40
10043211 High Binding Chimney Well Pack of 40
10680932 Medium Binding Chimney Well Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10577091 Supplier Greiner Bio-One Supplier No. 655061
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from crystal clear polystyrene for optimum visibility. Greiner BioOne™ Microlon™ Polystyrene 96-Well ELISA Microplates are tested in a colorimetric immunoasay for immunological research applications.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Tracebility from production through a defined LOT number system
  • Manufactured from crystal clear polystyrene
  • The footprint is compatible with automated systems
  • Well to well spacing : 9 mm
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Specifically treated to provide an incresed protein binding
  • Well profile: F-bottom / standard (flat bottom)
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Binding Property High Binding
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Product Type ELISA Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon 600
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Standard Well