missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Plate Sealing Mats, 7 mm Diameter, Round, Level 2 High-Throughput Applications

Product Code. 30286148 Shop All Thermo Scientific Products
Change view
Click to view available options
Pre-slit:
No
Yes
Material:
EVA
Silicone
Silicone Rubber
Silicone, PTFE
Format:
Domed Base
Flat Base
Flat Base, Large Penetration Area
Step Base, Small Penetration Area
No. per Pack:
100 Mats
5
5 Mats
50
Unit Size:
Pack of 100
Pack of 5
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pre-slit Material Format No. per Pack Unit Size
30286148 No Silicone Rubber Flat Base, Large Penetration Area 50 Pack of 50
15142739 No Silicone Step Base, Small Penetration Area 5 Mats Pack of 5
15332165 No Silicone Flat Base, Large Penetration Area 5 Pack of 5
15162739 Yes Silicone Step Base, Small Penetration Area 5 Mats Pack of 5
15152739 Yes Silicone Flat Base, Large Penetration Area 5 Mats Pack of 5
15122749 No Silicone, PTFE Flat Base 5 Mats Pack of 5
15132749 Yes Silicone, PTFE Flat Base 5 Mats Pack of 5
17473193 No EVA Domed Base 100 Mats Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 30286148 Supplier Thermo Scientific™ Supplier No. 60180M187
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use SureSTART™ WebSeal™ 96-Well Plate Sealing Mats, 7 mm Diameter, Round, Performance Level 2 for high-throughput chromatography analysis. They are specification certified and conform to ANSI-SLAS 4-2004 standards.

Thermo Scientific™ SureSTART™ Level 2 WebSeal™ 7 mm Diameter Round 96-Well Plate Sealing Mats are designed to ensure high quality data with an uninterrupted workflow for high-throughput applications using GC and HPLC/UHPLC and are ideal for regulated labs that use water and polar solvents.

These 96-well plate sealing mats are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards.

Performance Level 2 Mats – for High-Throughput Applications

  • Ideal for non-regulated labs using water and polar solvents
  • GC-MS and LC-MS tested
  • Suitable for HPLC/UHPLC,QA/QC, and sensitive applications
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, fluorescence, and ELSD
  • Eliminates cross contamination of samples
  • Resists coring and tearing
  • Superior resealability after multiple injections
  • WebSeal mats are manufactured from EVA (Ethylene-vinyl acetate) or pure silicone with or without a PTFE coating
  • Silicone mats are dry heat autoclavable with excellent chemical compatibility, and can withstand low temperatures down to -80°C

Reduce the risk of mis-injection – To seal your plates correctly, it’s important to use the right mat

  • Silicone Mats – Using a silicone mat with the correct profile reduces sample and solvent evaporation during your workflows, provides better consistency, and is ideal for long-term storage.
  • Pre-slit Mats – Pre-slit mats are effective at reducing mis-injections and can be used again for confirmatory runs on the same plates without solvent loss. Their slits allow the use of pipette tips as well as multichannel pipettes.
  • Adhesive Seals – Adhesive seals are effective as a one-time seal, but cannot be used for repeat runs. The exception is our three-layer re-sealing tape without glue at the cavity area.

Specification Certified 

  • All Thermo Scientific SureSTART WebSeal mats are dimensionally verified 

Lot Traceable

  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request 

About Thermo Scientific WebSeal Well Plates & Mats

Thermo Scientific WebSeal plates and mats are available in three performance levels:

  • Level 1 products are for everyday applications and are suitable for non-regulated labs using water and polar solvents
  • Level 2 products are for high-throughput applications and are suitable for regulated labs using water and polar solvents
  • Level 3 products are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Solvent Or Alcohol Resistant Alcohol Resistant
Material Silicone Rubber
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Pack 50
Plug Diameter 7 mm
For Use With (Application) HPLC, UHPLC, GC
Format Flat Base, Large Penetration Area
Pre-slit No
Product Line SureSTART WebSeal Level 2
Adhesive None
Pierceable Yes
Well Shape Round
Plate Compatibility 96-well Standard Plate
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.