missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Microplates for Fluorescence-based Assays

Product Code. 18832653
Change view
Click to view available options
No. of Wells:
96
384
Quantity:
10 Plates
30 Plates
Unit Size:
Pack of 10
Pack of 30
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Quantity Unit Size
18832653 384 10 Plates Pack of 10
15329413 96 10 Plates Pack of 10
30274967 96 30 Plates Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18832653 Supplier Invitrogen™ Supplier No. M33091
Estimated Shipment: 15-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.

Microplates for fluorescence-based assays are black-walled, clear-bottom microplates that minimize well-to-well crosstalk and autofluorescence. These 96- and 384-well microplates are compatible with standard fluorescence microplate readers and optimized for fluorescence-based applications including nucleic acid quantitation (e.g., Quant-iT assays, including PicoGreen, RiboGreen, and OliGreen).

Features of the microplates for fluorescence-based assays include:

  • Black-walled design—minimizes well-to-well crosstalk and autofluorescence
  • Clear-bottom wells—allows for optimal fluorescence detection
  • Compatibility—suitable for standard fluorescence microplate readers
  • Optimized for fluorescence microplate assays—ideal for nucleic acid quantitation (e.g., Quant-iT assays)
  • Versatile formats—available in 96- and 384-well configurations
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Content And Storage Store at room temperature.
Array 16 x 24
Autoclavable Not Autoclavable
Packaging 10-plates
Product Line Invitrogen
Color Clear Bottom, Black
Disposable Single-use
Product Type Microplates
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 40 μL
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
For Use With (Application) Nucleic Acid Quantification
Quantity 10 Plates
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.