missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Microsart™ Media

Product Code. 15932510 Shop All Sartorius Products
Change view
Click to view available options
Without Additives:
No Cholesterol
No Sucrose
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Without Additives Unit Size
15932510 No Cholesterol Pack of 100
15952510 No Sucrose Pack of 100
15942510 No Cholesterol Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15932510 Supplier Sartorius Supplier No. 1431347ACN
  • ONLINE EXCLUSIVE PRICE
    568.00 €
    500.00 € / Pack of 100
    Save 68.00 €
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enhance the safety of microbiological quality control in the pharmaceutical and biotech industries for microbial limits testing according to the USP and EP

  • Ready-to-use agar media dish (prefilled with different agar media types) with active lid
  • Sterile packaged and ready to use together with Microsart filter units
  • Active lid enables touch-free transfer of membrane, without using any tweezers and reduces the risk of secondary contamination
  • Liftable interior lid provides easy access to select colonies after incubation for further analysis
TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin
Form Gel
Product Type Growth Medium
Serum Level Serum-free
Sterility Sterile
Sterilization Method Gamma-irradiated
With Additives BPE
Without Additives No Cholesterol
For Use With (Application) Target microorganisms: total count
Formulation Solid Agar Medium
Culture Type Static Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.