missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Microsart™ Media

Product Code. 15952510 Shop All Sartorius Products
Change view
Click to view available options
Without Additives:
No Cholesterol
No Sucrose
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Without Additives Unit Size
15952510 No Sucrose Pack of 100
15932510 No Cholesterol Pack of 100
15942510 No Cholesterol Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15952510 Supplier Sartorius Supplier No. 1432247ACN
  • ONLINE EXCLUSIVE PRICE
    568.00 €
    500.00 € / Pack of 100
    Save 68.00 €
    11% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enhance the safety of microbiological quality control in the pharmaceutical and biotech industries for microbial limits testing according to the USP and EP

  • Ready-to-use agar media dish (prefilled with different agar media types) with active lid
  • Sterile packaged and ready to use together with Microsart filter units
  • Active lid enables touch-free transfer of membrane, without using any tweezers and reduces the risk of secondary contamination
  • Liftable interior lid provides easy access to select colonies after incubation for further analysis
TRUSTED_SUSTAINABILITY

Specifikationer

Classification Reduced Growth Factor, Very Low Protein (<90ng/mL)
Form Gel
Product Type Growth Medium
Serum Level Serum-free
Sterility Sterile
Sterilization Method Gamma-irradiated
With Additives Essential and nonessential amino acids
Without Additives No Sucrose
For Use With (Application) Target microorganisms: total count, stressed organisms found in pure water systems
Formulation Solid Agar Medium
Culture Type Bacterial Cell Culture
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.