missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Microtitration Plate
Click to view available options
Sterility:
Non-sterile
Sterile
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Unit Size:
Pack of 50
Description
- Multiple-well plates with 96 sample wells in clear PS.
- Flat bottom or V-bottom wells, single wrapped sterile.
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Binding Type | None |
| Color | Clear |
| Material | Polystyrene |
| Volume (Metric) Well | 0.404 mL |
| No. of Wells | 96 |
| Sterility | Sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction