Learn More
Scientific Industries SI™ Microtube Foam Insert
Product Code. 15577325 Shop All Ohaus Products
Change view
Click to view available options
No. of Positions:
60
Unit Size:
Each
Product Code. No. of Positions Unit Size
15577325 60 Each
1 options
Product Code. 15577325 Supplier Scientific Industries SI™ Supplier No. 83041418
  • ONLINE EXCLUSIVE PRICE
    76.75 €
    49.84 € / Pack of 1
    Save 26.91 €
    35% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Holds vessels in place while mixing. Scientific Industries SI™ Microtube Foam Insert fits snugly inside the Vortex-Genie 2 and Vortex-Genie Pulse 152mm platform attachment.
  • Accommodates up to 60 microtubes.
  • Fits into 152mm Platform attachment
  • Included in the Multiple Sample Starter Set
  • For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
TRUSTED_SUSTAINABILITY
Specifications
Material High-density foam
For Use With (Equipment) Vortex-Genie 2 family of mixers, Vortex-Genie Pulse
No. of Positions 60
Requires Platform Attachment 15577315