Learn More
Labnet™ Microtube Platform for VorTemp™ 56 Shaking Incubator
Product Code. 15103883
Change view
Click to view available options
For Use With (Equipment):
VorTemp™ 56 Shaking Incubator
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15103883 VorTemp™ 56 Shaking Incubator Each
1 options
Product Code. 15103883 Supplier Labnet™ Supplier No. S2056R
  • ONLINE EXCLUSIVE PRICE
    206.85 €
    189.48 € / Pack of 1
    Save 17.37 €
    8% Off
Estimated Shipment: 04-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
VorTemp™ 56 Shaking Incubator Microtube Platform is intended for shaking 56 x 1.5/2 mL test tubes.
  • Designed for simultaneous heating and mixing of small samples, the VorTemp™ 56 microplate shaking incubator features interchangeable platforms for microtubes and microplates. Both platforms are supplied with the unit.
  • Temperature in the VorTemp™ 56 microplate shaking incubator is adjustable over a broad range, making the unit useful for a variety of applications, including bacterial cultures in tubes and microplates. Mechanical convection provides a uniform and stable environment within the chamber. The VorTemp™ 56 shaking incubator may also be used in the cold room for sub-ambient applications.
    • Shake and incubate samples simultaneously
    • Complete with platforms for microtubes and microplates
    • Intelligent control
    • Broad temperature and speed range
  • The Microtube Platform is intended for shaking 56 x 1.5/2 mL test tubes. There are accessory adapters available which allow for running several different microtube sizes. In addition, the platform and adapters are autoclavable.
Specifications
Holds 56 x 1.5/2 mL Test Tubes
Product Type Shaker Platform
For Use With (Application) Shaking Test Tubes
For Use With (Equipment) VorTemp™ 56 Shaking Incubator