missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICU1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142722
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize reported isoforms 1 and 2 (NP_006068.2; NP_001182447.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
MICU1 is a key regulator of mitochondrial calcium uptake required for calcium entry into mitochondrion. It may act as a calcium sensor via its EF-hand domains, gating the activity of a calcium channel partner. MICU1 induces T-helper 1-mediated autoreactivity, which is accompanied by the release of IFNG.
Specifications
| MICU1 | |
| Polyclonal | |
| Unconjugated | |
| MICU1 | |
| ara CALC; atopy-related autoantigen CALC; C730016L05Rik; CALC; calcium binding atopy-related autoantigen 1; calcium uptake protein 1, mitochondrial; calcium-binding atopy-like autoantigen 1; calcium-binding atopy-related autoantigen 1; Calcium-binding atopy-related autoantigen 1 homolog; calcium-binding atopy-related autoantigen-like protein; cbara1; DKFZp564C246; EFHA3; FLJ12684; Hom s 4; MICU1; mitochondrial calcium uptake 1; MPXPS; si:ch211-199i24.4; zgc:162148 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10367 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BPX6 | |
| MICU1 | |
| Peptide with sequence C-EYENRIRAYSTPDK (aa 113-126). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion