missing translation for 'onlineSavingsMsg'
Learn More

Midecamycin, MedChemExpress

Product Code. 16468826
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468826 100 mg 100mg
16464637 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468826 Supplier Medchem Express Supplier No. HYB1908
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Midecamycin, an acetoxy-substituted macrolide antibiotic, is tested against gram-positive and gram-negative bacteria.

Midecamycin, an acetoxy-substituted macrolide antibiotic, is tested against gram-positive and gram-negative bacteria.

Specifications

Chemical Name or Material Midecamycin
Physical Form Solid
CAS 35457-80-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C41H67NO15
For Use With (Application) COVID-19-immunoregulation
Synonym SF-837 Antibiotic SF-837
Solubility Information DMSO : ≥ 36 mg/mL (44.23 mM)
SMILES O[C@H]1[C@](O[C@H](C)[C@@H](O[C@@](O[C@@H](C)[C@@H]2OC(CC)=O)([H])C[C@]2(O)C)[C@@H]1N(C)C)([H])O[C@@H]([C@H](C[C@H]([C@H](/C=C/C=C/C3)O)C)CC=O)[C@H]([C@](CC(O[C@@H]3C)=O)([H])OC(CC)=O)OC
Molecular Weight (g/mol) 813.97
Formula Weight 813.97
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.