missing translation for 'onlineSavingsMsg'
Learn More

Milnacipran hydrochloride, MedChemExpress

Product Code. 16478146
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478146 100 mg 100mg
16474267 50 mg 50mg
16468378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478146 Supplier Medchem Express Supplier No. HYB0168A
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.

Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.

Specifications

Chemical Name or Material Milnacipran hydrochloride
Physical Form Solid
CAS 101152-94-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H23ClN2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : ≥ 100 mg/mL (353.59 mM) ∣DMSO : ≥ 48 mg/mL (169.73 mM)
SMILES O=C([C@@]1(C2=CC=CC=C2)[C@@H](CN)C1)N(CC)CC.Cl
Molecular Weight (g/mol) 282.81
Formula Weight 282.81
Percent Purity 99.92%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.