missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mimecan Polyclonal Antibody

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148614

Product Code. 30533772

  • 479.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta. Mimecan is a small proteoglycan which contains tandem leucine-rich repeats. Mimecan is encoded by the OGN gene, a member of the small leucine-rich proteoglycan (SLRP) family of proteins. Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta (TGF-beta-1 or TGF-beta-2) and may regulate osteoblast differentiation. High expression of mimecan protein may be associated with elevated heart left ventricular mass. Alternative splicing results in multiple transcript variants of mimecan. Alternatively spliced transcript variants that encode mimecan have been found.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mimecan
Polyclonal
Unconjugated
OGN
3110079A16Rik; corneal keratan sulfate proteoglycan; mimecan; mimecan proteoglycan; mimican; OG; Ogn; OIF; Osteoglycin; osteoglycin/mimecan; osteoinductive factor; SLRR3A
Goat
Antigen affinity chromatography
RUO
4969
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
P20774
OGN
Mouse myeloma cell line NS0-derived recombinant human Mimecan, Ala20-Phe298.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.